Weller® Zero Smog® Fume Extractor, Apex
Supplier: Cooper Tools
ZEROSMOGELKIT1N
76249-450EA 2570.57 USD
76249-450
Weller® Zero Smog® Fume Extractor, Apex
Fume Extractors
This fume extraction unit purifies air at up to two workplaces.
  • Easy filter exchange
  • Quick and simple installation
  • Portable fume extraction unit
Particle filter separation is >99.95%. Has a blower vacuum of 3000 Pa and blower capacity of 220 m3/h.
Applications include hand soldering, heavy duty soldering (solder bath, selective wave), gluing, cleaning, filling, finedust.
Delivery information: Includes Zero Smog® 2 fume extraction unit, 120 V H13 HEPA filter (10% active carbon foam), low gas M5 Medium dust pre-filter and ALFA funnel.