Weller® WX Series Solder System, Apex
Supplier: Cooper Tools
T0052920299N
76249-358EA 1654.46 USD
76249-358
Weller® WX Series Solder System, Apex
Soldering Supplies Soldering Irons
WX series solder system from features a glass touch screen, intuitive user interface, energy-saving sleep mode, and multi-language display.
  • The glass capacitive tough screen has a patented anti-static, chemical and temperature resistant coating
  • Voltage: 110 V
A multi-purpose USB port facilitates easy firmware updates, parameter setting, monitoring, and personalization. The multi-language menu is accessed via a turn and click control wheel with finger guide and enter key. An integrated sensor system, customized bit calibration independent of the base station, a built-in signal reader, and internal memory.
A wide selection of soldering tips and other soldering accessories, such as fume extraction systems and heating plates, complete the system for a total soldering solution.
Certifications: RoHS complaint.