Drill Monster® Jobbers Length Twist Drills, Cutting Tool Supply
Supplier: CUTTING TOOL SUPPLY TE
450-301890 450-301610 450-101875 450-101562 450-202570 450-203970 450-101250 450-300670A 450-301040 450-301285 450-300550
76249-244EA 54.19 USD
76249-244 76249-246 76249-248 76249-250 76250-096 76250-100 76250-102 76250-684 76250-932 76251-334 76252-800
Drill Monster® Jobbers Length Twist Drills, Cutting Tool Supply
Tool Sets
These general-purpose jobber length twist drills are designed in a manner to assure true, accurate and smooth holes.
  • 118°- drill point style
  • 25° helix - RHS/RHC (Right Hand Cut/Right Hand Spiral)
  • Diameter Tolerance: +0.0000/−0.0005"
  • Available in fractional, letter and wire gage size
These drills are made of solid carbide (C-2 grade premium sub-micron carbide with 10% cobalt), which provides higher wear resistance.
Ideal for use on non-ferrous alloys, cast iron, plastics, aluminum, and highly abrasive materials.
Ordering information: Available as either with or without coating (TiN or TiAlN coated).