Morse® 2-Flute Miniature Long Length Double End Mills, Cutting Tool Supply
Supplier: CUTTING TOOL SUPPLY TE
43253-MO
76249-142EA 96.68 USD
76249-142
Morse® 2-Flute Miniature Long Length Double End Mills, Cutting Tool Supply
Tool Sets
These miniature center cutting shank end mills are designed for small diameter milling of slots, keyways and pockets.
  • Shorter length provides maximum rigidity
Center cutting end allows for plunge cutting like a drill into solid material.