Momentive™ Electronics Grade Silicone Adhesive Sealant, HISCO
Supplier: Hisco
RTV162-300ML
76249-048EA 228.96 USD
76249-048
Momentive™ Electronics Grade Silicone Adhesive Sealant, HISCO
Adhesives
This is a non-corrosive and cures to a tough, resilient silicone rubber at room temperature.
  • Alcohol like odor
  • Hardness: 35 Shore A
This is compatible with metals glass ceramic silicone rubber some rigid plastics.
Certifications: MIL-A-46146B compliant.