Self-Laminating Labels, Brady
Supplier: Brady Worldwide
29766
76249-046PK 1490.73 USD
76249-046
Self-Laminating Labels, Brady
Labels
Self-laminating Labels helps in smudge resistance, solvent resistance and elevated temperature performance.
  • Chemical, temperature and corrosion resistant
  • Humidity, Smear and UV resistant
Simple to customize using laser print technology. Rectangle in shape.
Service temperature: −94 °F to +230 °F (−70 °C to +110 °C).