DME-1A Four Tools Control Unit, JBC
Supplier: JBC TOOLS USA INC. TE
DME-1A
76248-702EA 1904.29 USD
76248-702
DME-1A Four Tools Control Unit, JBC
Soldering Supplies Soldering Stations
The DME manages up to 4 tools simultaneously and it is fully compatible with most of tools.
  • The easy-to-use menu helps you monitor the work in real time
  • Operating temperature: 10 to 40 ºC
  • RJ12 is act as both PS and robot connector
The DME also provides with extra applications that support work such as USB microscope, file storage, unit converter and video.