Conductive Box with Foam, Menda
Supplier: Protektive Pak
57000
76248-506EA 9.46 USD
76248-506
Conductive Box with Foam, Menda
Boxes ESD-Safe Boxes
This provides ESD protective storage and handling of ESD sensitive items.
  • RTT <1×10 E5 ohms - electrostatic charges able to be removed when grounded
  • Secure snap closure keeps boxes from accidentally opening
  • Conductive foam inserts provide protection from vibration and shock
ESD protective symbol molded into the cover of the box as per per ESD S541.