Depth Micrometer, Sid Tool Co
Supplier: SID TOOL CO INC TE
87130597
76248-458EA 2273.77 USD
76248-458
Depth Micrometer, Sid Tool Co
Micrometers
This has interchangeable rods (Ø 4 mm), with lapped measuring end, provide a wide measuring range, The rod length can be adjusted in 1" or 25 mm increments.
  • Ratchet stop for constant force
  • Measuring rod clamp.
  • SPC output
Packaging: Fitted plastic case