Soldering Tool Control Units, JBC
Supplier: JBC TOOLS USA INC. TE
DI-1D
76248-382EA 2196.25 USD
76248-382
Soldering Tool Control Units, JBC
Soldering Supplies Soldering Stations
These control units manages for soldering or desoldering.
The 2 tools control unit manages up to 2 tools simultaneously, it works with the exclusive heating system and the intelligent sleep and hibernation modes, with benefits such as station customization, full control of soldering processes and remote work management. The easy-to-use menu offers innovative functions to help you monitor the work in real time: graphics, partial counters or tool port monitoring.
The 1 tool control unit manages 1 tool. The DI works with tne exclusive heating system and the intelligent sleep & hibernation features to extend a tip's life up to 5 times longer.Using the menu you can personalize over 20 parameters to help manage the soldering process. The USB connector allows to update software, create graphs and manage parameters from a PC.
For a basic working system, need at least 1 stand, 1 tool and 1 cartridge.