Protective Film Tape, 3M, Tapecase
Supplier: TAPECASE LTD. TE
31825C-10X300F
76248-366EA 209.12 USD
76248-366
Protective Film Tape, 3M, Tapecase
Seals General Purpose Sealing Films
This is made of low density polyethylene that has a low-tack acrylic adhesive system to help provide temporary protection of mill finished stainless steel and aluminum coils.