Tacki-Pak™ Component Box, Conductive Containers
Supplier: CONDUCTIVE CONTAINERS INC. TE
TP3545
76248-308EA 30.26 USD
76248-308
Tacki-Pak™ Component Box, Conductive Containers
Boxes ESD-Safe Boxes
Tacki-Pak™ provides an ESD-Safe, non-adhesive, tacky surface that will limit component movement. No longer will parts slide into each other causing damage.
  • Tack level I rating range 2.0 - 4.0 (rating is in foot pounds of force)
  • Box surface resistance 103- 106 ohms
  • Tacki surface resistance 105 - 109 ohms
  • Reusable - clean with IPA (wipes) to remove FOD and rejuvenate surface tack level
This is a flat, user friendly surface that minimizes residue transfer to the component. It has stable, reusable and cleanable ESD-Safe properties.