Opto-Wipes™ Precision Lens Cleaning Wipes, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71755-01 71755-02 71755-04 71755-03 LCW-304
100489-358PK 64.19 USD
100489-358 100489-360 100489-364 100489-362 76248-124
Opto-Wipes™ Precision Lens Cleaning Wipes, Electron Microscopy Sciences
Wipes Disposable Wipes and Towels Lens Papers
A strong hydro-entangled mixture of polyester and cellulose fiber; designed for clean room environments.
  • This special material offers softness, is lint-free, and traps particles and residue quite easily
Wipes made of super-soft fibers prevent the scratching of precision surfaces and its strong fiber matrix maintains integrity when wet. Opto-Wipes™ are highly absorbent which retains and withstands solvents such as MEK, methylene chloride, acetone, and alcohol. Its efficient particle trapping matrix lifts particles and residue away from the surface. They are washable and reusable.