HumiSeal® 1A33 Urethane Conformal Coating, Ellsworth Adhesives
Supplier: ELLSWORTH ADHESIVES CUST SERV TE
1A33
76248-030EA 94.65 USD
76248-030
HumiSeal® 1A33 Urethane Conformal Coating, Ellsworth Adhesives
Sprays
HumiSeal® Urethane conformal coating is a one component, polyurethane coating that is used for circuit board applications.
  • Well suited for general printed circuit board applications
  • Specific ravity of 0.95
  • Flash point of –0.1 °C
It contains no free isocyanates and fluoresces under UV light to aid inspection. It can be applied by spraying, brushing, or dipping methods.