Sticklers® CleanClicker™ End-Face Cleaners, Microcare
Supplier: MicroCare
MCC-CCU125
76247-912EA 435.8 USD
76247-912 76247-914 76252-936
Sticklers® CleanClicker™ End-Face Cleaners, Microcare
Wipes Cleaning Kits and Pads
The Sticklers® CleanClicker™ 1.25 mm and 2.5 mm fiber optic connector cleaners are the latest cleaning innovation from the experts at Sticklers. These rugged, precision cleaners use a smooth click-to-clean push-action with light tip forces to reduce the risk of end-face damage.
  • Easy-to-use on outside plant and premise networks
  • Part of the industry’s most complete line of fiber optic cleaning products
  • Color coded labels and packaging minimize user error and speed cleaning
  • Cleans dust, particulate and oil contamination
With over 750 cleans, the Sticklers CleanClicker is perfect for cleaning any size network, any installation, from single to thousands of connectors.
Sticklers CleanClicker cleaners are packaged color-coded byconnector size to help users quickly ensure they have the right cleaner for the connector at hand.