Antistatic Utility Tape Dispenser 620, with base, 3M™
Supplier: 3M
7100049758
76247-892EA 607.81 USD
76247-892
Antistatic Utility Tape Dispenser 620, with base, 3M™
Label and Tape Dispensers
Tape dispenser 620 is designed for use in ESD protected areas. The dispenser is made of conductive and static-dissipative materials which will minimize or eliminate problems associated with conventional dispensers.
  • Minimizes problems with static-dissipative and conductive material construction
  • Minimizes floor space requirements
Stainless-steel blade easily cuts the tape with maximum strength.