Pan-Ty® Cable Ties, Panduit
Supplier: PANDUIT TE
PLT3I-C PLT.6SM-M0
76242-662PK 44.22 USD
76242-662 76243-242 76243-246 76245-000 76245-204 76247-794
Pan-Ty® Cable Ties, Panduit
Ties
Versatile cable ties can be used in countless applications. One-piece construction for consistent performance and reliability. Lowest threading force of any one-piece cable tie in the industry.
  • For indoor use
  • Operating temperature: –60 to 85 °C
Curved tip is easy to pick up from flat surfaces and allows faster initial threading to speed installation.
Certifications: UL Listed for use in plenum or air handling spaces per NEC except PLT.6SM.