Self Fusing Tape, Grainger
Supplier: GRAINGER
GL20B67100
76247-762EA 69.79 USD
76247-762
Self Fusing Tape, Grainger
Tapes
Use this adhesive-free, protective overwrap to help create a permanent seal.
  • Liquid pressure range: 0 to 700 psi
Triangle self fusing tape features a triangular shape that helps assure a smooth and uniform wrap over difficult shapes such as bus bars, cable harnesses, coils, and splices. Ideal for use in insulating electrical connections, protecting pipe threads, friction wrap for handle grips, tension wrap for bundles of wires and hoses, temporary hose repairs, and corrosion protection.