ClearView™ Standard Wall Closed End Tubing, TRC Series, Caplugs
Supplier: Caplugs
TRC-625-014-7.000
76247-684PK 2905.16 USD
76247-684
ClearView™ Standard Wall Closed End Tubing, TRC Series, Caplugs
Tubing Transfer Tubing
Series TRC heavy wall ClearView™ Tubing has a clear closed end. Heavy wall thickness adds extra protection and durability.
  • Made of PETG
  • Color: Natural (clear)
Available in a variety of lengths and ranges from 0.5 to 3.57" diameter to meet all of the packaging needs.