Desiccant in Tyvek® Pouch, SCS Division of Desco
Supplier: Desco
2PLDES150
76247-632CS 316.2 USD
76247-632
Desiccant in Tyvek® Pouch, SCS Division of Desco
Desiccators Desiccants
These dessicants provide protection for moisture sensitive electronics (such as SMT – Surface Mount Technology).
  • Secured in strong, sulphur-free non-woven plastic pouches
  • Can be baked and re-used
  • No hazardous substances
  • Recommended for use in clean rooms
Recommended amount of desiccant is dependent on the interior surface area of the bag to be used.