Fume Extraction System, FA-430, American Hakko Products
Supplier: AMERICAN HAKKO PRODUCTS INC TE
FA430-16
76247-384EA 2223.4 USD
76247-384
Fume Extraction System, FA-430, American Hakko Products
Fume Extractors
FA-430 fume extractor is specially designed for use at soldering stations by extracting the contaminated air directly at the source.
  • Powerful suction
  • Three airflow adjustment modes: High, Medium, Low
  • Free standing ducts
Contaminated air is captured from the workplace surrounding the soldering station by the hood or hoods, and transferred to the FA-430 unit by rigid ducts and a special replacement filter notification function notifies the operator when the filter needs to be replaced.
Three stages of filtering remove the contaminants from the air. The purified air is then returned to the area where the FA-430 is located
The pre-filter removes coarse particles, such as dust, thus extending the life of the main filter. The main filter provides the greatest amount of surface area within the volume of the filter case. The standard filters contain an activated carbon element built into the filter. This filter is designed to last the life of the main filter.
To maintain optimum filtering efficiency, special filter replacement indicators on the front panel notify you when it is time to replace the filters.
Ordering information: Loc-Line duct with rectangular nozzle sold separately.