Shelving Storage Systems, Midwest Container Systems
Supplier: MIDWEST CONTAINER SYSTEMS LLC TE
J1836C
76247-332EA 133.09 USD
76247-332
Shelving Storage Systems, Midwest Container Systems
Furniture Shelves
Olympic wire shelving made of carbon-steel will exceed all your storage needs. Open construction allows use of maximum storage space of cube. Chrome and black (certain sizes) finishes are perfect for retail applications.
  • Open wire design that minimizes dust accumulation and allows a free circulation of air
  • Greater visibility of stored items and greater light penetration
  • Can be loaded/unloaded from all sides
  • Wire shelving that can change as quickly as your needs change
  • Shelf wires run front to back allowing for items to slide on and off shelves smoothly
  • Shelves can be adjusted at 1" intervals along entire length of post
Green epoxy finish with chromat substrate is rust resistant and is suitable in cold and/or wet environments.
Certifications: NSF Approved.
Delivery information: Each shelf includes split-sleeves to attach it to posts.