Tyvek® Bag Desiccants, Uline
Supplier: ULINE
S-1605
76247-240EA 832.48 USD
76247-240
Tyvek® Bag Desiccants, Uline
Desiccators Desiccants
Tyvek® provides a tear-proof, dust-free packet.
The Uline desiccant pack protects up to 6.67 cu. ft. Recommended for shipping pharmaceuticals, electronic components and food.
Certifications: Meets FDA specifications for contact with food and drugs.