Omega Plus Vacuum Cleaner, VACOMEGA, Jensen Tools
Supplier: TESTEQUITY LLC TE
VACOMEGA
76247-142EA 778.12 USD
76247-142
Omega Plus Vacuum Cleaner, VACOMEGA, Jensen Tools
Vacuum Cleaners
This Omega Plus vacuum cleaner is an excellent and economical immediate containment option for cleaning static sensitive equipment such as computers, copiers, printers, data centers, ATM’s, gaming, banking, robotic and telecom applications.
  • ESD-safe
  • Made from high impact diamond ABS Polymer for durability and extended life
  • Lightweight and easy to use
  • Can be operated with gloved hands and carried around on unsteady surfaces, like ladders
  • Optional ULPA Filter Cartridge - 99.999% efficient at 1.2 µm
This vacuum features ultrafine filtration, self-storage and contained accessories, 1000 hour rated Ametek® Advantek II motor and 10' detachable power cord. The one gallon ultrafine filter has 43 pleat cellulose media and is 88% efficient at 0.3 µm or larger.
The ultrafine filter can capture toner, color toner, dust and debris. The vacuum and accessories are ESD safe providing a static path to ground. The tool box structure and self-contained accessories makes this vacuum convenient for mobile service technicians.
Delivery information: Includes vacuum with 1 HP motor, self-storing ESD-safe accessories, 6' ESD safe toner proof hose, 16" ESD safe rubber flex neck tool, ESD-safe crevice tool with brush, detachable power cord, 0.3 micron disposable filter canister.