Henkel Loctite 222 Threadlocker Adhesive, Ellsworth Adhesives
Supplier: ELLSWORTH ADHESIVES CUST SERV TE
231125
76247-136EA 56.77 USD
76247-136
Henkel Loctite 222 Threadlocker Adhesive, Ellsworth Adhesives
Adhesives
This is a one component anaerobic material designed for the locking and sealing of threaded fasteners which require easy disassembly with standard hand tools.
  • Dimethacrylate ester
  • Cure time up to 72 hours depending on the substrate used, the bondline gap, and the temperature.
Used for applications such as adjustment of set screws, small diameter or long engagement length fasteners where easy disassembly is required without shearing the screw.