Splice Clip Frame, Splicetronics Solutions
Supplier: SPLICETRONICS SOLUTIONS INC TE
CP408
76247-080BX 212.13 USD
76247-080
Splice Clip Frame, Splicetronics Solutions
Tool Sets
Bandolier type clip/shim used with the ST410 splice tool and tape of any width.
  • Guarantees a secure connection of SMT component reels
This frame increase SMT pick and place machine output by 5 to 30%.
Packaging: 20 clips per frame.