Tape, Double Splice, Splicetronics Solutions
Supplier: SPLICETRONICS SOLUTIONS INC TE
DSU308
76247-078BX 191.41 USD
76247-078
Tape, Double Splice, Splicetronics Solutions
Tapes
Double splice tape designed with eight holes for accurate alignment. Possible to join SMT component reels without the splice tool and clip. Increase SMT pick and place machine output by 5 to 30%.