Electronic Tape Dispenser, Jensen Tools
Supplier: TESTEQUITY LLC TE
ZCM1000
76247-016EA 2448.9 USD
76247-016
Electronic Tape Dispenser, Jensen Tools
Label and Tape Dispensers
Ideal for medium to high production of repetitive taping for industrial applications.
  • Photocell actuated feed and cut or push-button feed and cut
  • LED digital length adjustment
  • Adjustable pressure roller for various thicknesses of tape
  • Blades good for up to 200000 cuts (dependent on type of material)
  • Comes with a 3-in-1 core: 1, 1.4, 3"
  • Accuracy of tape length is ±0.04" dependent of the type of material
Accepts tape widths from 0.28 to 2" and cuts tape lengths from 0.79 to 39". Program a defined length and automatically advance and cut tape.