Aerostat® Ionizing Blower, Simco Ion
Supplier: Simco
4011009
76246-960EA 4245.91 USD
76246-960
Aerostat® Ionizing Blower, Simco Ion
Ionizers
Aerostat FPD four fan ionizing blower is designed to control electrostatic charges in assembly and electronics manufacturing.
  • Extremely fast static discharge
  • Built-in emitter point cleaner for simplified maintenance
  • Ion balance and ion output LED provide real-time ionization status
  • Emitter Points never need replacement
The Aerostat FPD Ionizing Air Blower provides superior fast neutralization over an extended surface area. Available in two, three and four-fan models, this high velocity overhead blower provides superior coverage up to 5'.