Diamond Wheel Cutter, Dremel
Supplier: DREMEL SERVICES INC. TE
545
76246-918EA 84.03 USD
76246-918
Diamond Wheel Cutter, Dremel
Cutters
Diamond wheel is completely coated with fine diamond particles for working with hard materials. Designed to be long lasting and not lose its shape or break, it provides smooth, fine cuts.
  • Not recommended for metal
Ideal for cutting, sawing, carving, sharpening and filing hard materials-including marble concrete, brick, porcelain, ceramics, epoxy and hard woods.
Caution: Do not run in excess of 20000 rpm.