Polyester Overlaminate Labels, WorkHorse®, Brady
Supplier: Brady Worldwide
16101
76241-584EA 296.8 USD
76241-584 76241-588 76246-794
Polyester Overlaminate Labels, WorkHorse®, Brady
Labels
General purpose overlaminate that withstands high temperatures, dirt, oil and cleaning solvents while displaying excellent clarity.
  • Ideal for use on PC boards
  • Chemical and corrosion resistant
  • Humidity, UV and weather resistant
The Die-Cut overlaminate is made of clear polyester material.