Tip Test Probe Kit with Slim Retractable Probe, Newark/Element
Supplier: NEWARK/ELEMENT 14 TE
5953A
76246-758EA 142.08 USD
76246-758
Tip Test Probe Kit with Slim Retractable Probe, Newark/Element
Cables
Test probe kit with slim retractable probe is fully insulated to within 1/8" of tip. Needle point tip adjusts to desired length from 0 to 3". Stainless steel needle sharp point easily pierces varnish and thin insulation.
  • 30 VAC/60 VDC Maximum operating voltage
  • Flexible at low temperature
Tip can be re-pointed if necessary. Multi-stacking banana plug molded directly to the wire. 4' test lead wire is silicone insulated for high temperature resistance.