Soldering Tip Chisel, OK International
Supplier: OK INTERNATIONAL TE
SFV-CH25AR
76246-684EA 45.1 USD
76246-684
Soldering Tip Chisel, OK International
Soldering Supplies Soldering Tips
Chisel Original Easy Access Geometry.
The SxV Tips are for use with the soldering tip hand-piece of the PS and MFR Platforms. Recommended for heavy duty applications.