Soldering and Rework Cartridge, OK International
Supplier: OK INTERNATIONAL TE
SFP-CHL20
76246-376EA 81.09 USD
76246-376
Soldering and Rework Cartridge, OK International
Soldering Supplies Soldering Tips
The SxP Cartridges are for use with the MFR-H1-SC soldering cartridge hand-piece of the MFR Single and Dual Platforms (MFR-1100 and MFR-2200). They provide a wide range of options addressing specialized soldering and rework applications. The second digit denotes substrate material.