Hot Mill Plus Kevlar® Gloves, Plastic Process Equipment®
Supplier: PLASTIC PROCESS EQUIPMENT INC TE
HML-650
76246-134PK 163.14 USD
76246-134 76246-330
Hot Mill Plus Kevlar® Gloves, Plastic Process Equipment®
Gloves
Designed to withstand heat up to 500 °F, plus superior cut resistance.
  • Resists abrasions, cushions hands
  • No seams for maximum dexterity
  • Contains no asbestos
The heat resistant inner layer is bonded to the Kevlar® blended outer layer, combining protection from cuts and heat. The outer surface is coated with special heat resistant nitrile rubber that improves grip and won't melt.