ESD Dissipative Turntable, Menda
Supplier: Protektive Pak
47200
76246-222EA 329.16 USD
76246-222
ESD Dissipative Turntable, Menda
Furniture Shelves
Ideal for working on heavier assemblies in an EPA.
  • ESD protective for use with ESD sensitive assemblies
  • Durable 1/4" thick dissipative, crosslink polyethylene foam
  • Galvanized steel turntable located at the bottom
Volume resistance 1×106 to <1×109 ohms per ANSI/STM11.12. Electrostatic charges removed to ground via contact on grounded ESD worksurface or via ground cord from ring terminal.
Certifications: Meets worksurface required limit of ANSI/ESD S20.20 per ANSI/ESD S4.1 and ESD TR53.