Strap for Wrenches, Newark/Element
Supplier: NEWARK/ELEMENT 14 TE
TG70-18
76246-160EA 612.33 USD
76246-160
Strap for Wrenches, Newark/Element
Wrenches
This wrench is made of the following materials.
  • Wrench handle - aluminum alloy/nickel plate
  • Wedge - stainless steel/passivated
  • Strap - impregnated fabric
Straps are 12.7 mm (1/2") in width. Double wrap for heavy duty range.