PowerLock® Pocket Tape Measurers, Jensen Tools
Supplier: TESTEQUITY LLC TE
33-115
76246-128EA 29.39 USD
76246-128
PowerLock® Pocket Tape Measurers, Jensen Tools
Measuring Tapes
Back of blade provides scale that measures up to 3' diameters.
  • Polymer-coated blade withstands abrasion for long life
  • Chrome high-impact ABS case is job site tough
  • Slim case fits easily in pocket
  • Secure blade lock won't creep during measurements
Corrosion-resistant Tru-zero end hook for accurate measurements.