Heavy Duty Soldering Iron Kit, American Hakko Products
Supplier: AMERICAN HAKKO PRODUCTS INC TE
FM2030-02
76246-104EA 612.95 USD
76246-104
Heavy Duty Soldering Iron Kit, American Hakko Products
Soldering Supplies Soldering Irons
Heavy Duty Soldering iron Kit is the ideal companion for the Hakko FM-203 soldering station or the FM-206 rework station.
  • Compatible T22 tips
  • Soldering Iron With Holder
Provides a higher mass tip with double the power of conventional T15 tips. This iron is ideal for soldering difficult-to-solder heavy ground planes and/or high heat sink applications.
Ordering information: T22 tips has to be ordered separately.
Delivery information: Supplied with holder (with sleep functionality), tip cleaner and other affiliated parts.