Crescent® First Generation Adjustable Wrenches, Apex
Supplier: Cooper Tools
CRES-AC18C
76245-882EA 59.39 USD
76245-882
Crescent® First Generation Adjustable Wrenches, Apex
Wrenches
These first-generation adjustable wrenches are ideal for professional, industrial or home use.
  • Tension spring stabilizes jaw and knurl adjusts easily
  • Alloy steel and heat treated
  • Polished Head
  • Chrome plated finish resists rust and corrosion
The wide capacity jaw allows each tool to work with more fastener sizes and wider handle ensures more comfort and less stress to the hand. Proportioned dimensions for greatest strength and minimum weight.