Fume Extractor Arms, BVX, OK International
Supplier: OK INTERNATIONAL TE
BVX-ARML
76245-838EA 406.14 USD
76245-838
Fume Extractor Arms, BVX, OK International
Fume Extractors
This fume extractor arms are an economic solution while providing good airflow rates and high flexibility. The spiral rolled duct can be tightened in stiffness and can be positioned precisely.
  • Air flow rating of 75 m3/h (45 cfm)
  • Ducting with 40 mm (1,75") nozzle
  • BVX Arms compatible with all units