Tape Dispenser, Stainless Steel, Ultratape Industries
Supplier: Ultratape Industries
UT962
76245-618EA 713.36 USD
76245-618
Tape Dispenser, Stainless Steel, Ultratape Industries
Label and Tape Dispensers
The Dispenser is made from industrial 304 grade stainless steel with a mounting bracket on the bottom.
  • Corrosion resistant
  • Replaceable serrated blade is electroplated chrome buffed to a dull shine
Designed to hold one roll, with a 3" core, up to 2" wide and 220 yards in length.