Self Adhering Safety Tape, Magid Glove
Supplier: Magid Glove
41308-3/4
76245-586PK 121.54 USD
76245-586
Self Adhering Safety Tape, Magid Glove
Tapes
Guard-Tex self adhering safety tape is the wrap-around tape that guards hands, fingers and wrists from rough, sharp or hot surfaces.
  • Protects against cuts, abrasions and minor burns
  • Sticks only to itself, not to hair, skin or clothes
It works where gloves won't. You can maintain your sense of touch, dexterity and freedom. This tape is perfect for working in practically every manufacturing operation and handling components of all sizes.
It protects against contamination and prevents fingertips and accidental etching. Tape sticks only to itself, not to skin, hair, clothing or tools; making it easy to apply an painless to remove.