3M™ Scotch-Weld™ Metal Primer
Supplier: 3M
7000000907
76245-582CS 1778.45 USD
76245-582
3M™ Scotch-Weld™ Metal Primer
Adhesives
3M™ Scotch-Weld™ metal primer is a one-component, metal and glass primer designed for applications using 3M™ Scotch-Weld™ film and liquid structural adhesives.
  • Improves glass and metal adhesion
  • Resistance to environmental exposure for epoxy and urethane structural adhesives
  • Ensures complete wetting of film adhesive to adhered surfaces
  • Easy application and quick drying
Formulated to improve metal and glass adhesion, this film and liquid adhesive primer simplifies production scheduling by protecting the cleaned surfaces until the bonding operations can be completed.