Scotchcode Wire Marker Tape Dispenser, 3M™
Supplier: 3M
7000058253
76245-442CS 949.01 USD
76245-442
Scotchcode Wire Marker Tape Dispenser, 3M™
Label and Tape Dispensers
This tape is ideal for marking wires and cables and has pre-printed numbers and a high-tack, acrylic adhesive to ensure firm adhesion.
  • Refillable dispenser
  • Compact design for easy handling
  • Includes 1 each of 0 to 9 numbered tape rolls
  • Features an end loop to easily suspend it from a tool pouch or belt
It has a compact design for easy handling and 10 refillable compartments to hold non-smear polyester tape rolls. This carry-along dispenser has an end loop that hooks it easily to a tool pouch/belt for added convenience. The pre-printed tapes have a high-tack, acrylic adhesive to resist solvents, oil and water while adhering firmly to insulation surfaces.
Certifications: RoHS 2011/65/EU Compliant.