Lead Extractor, Jensen Tools
Supplier: TESTEQUITY LLC TE
81-1-45
76245-424EA 155.65 USD
76245-424
Lead Extractor, Jensen Tools
Puller and Inserter Tools
Shielded lead wire extractor ideal for removal of insulated lead wire from braided shield.
  • Eliminates broken shield strands
  • Production increases to 800%
  • Color coded for quick identification
  • Leaves braid intact for easy, neat soldering
Similar to a hypodermic needle, the lead extractor ejects the lead wire from a breakout in the shield produced by the tube point.