Foot Valve Dispenser, OK International
Supplier: OK INTERNATIONAL TE
TS924V
76245-410EA 2042.27 USD
76245-410
Foot Valve Dispenser, OK International
Fluid Adhesive Dispensers and Pistons
This dispenser offers increased production combined with dispenser economy.
  • Ideal for applications where electricity is prohibited
  • Robust design
  • Floor mounted conserves bench space
  • No programming required
Features a built in pressure regulator, pressure gauge and a unique fast-dump, three-way air valve. Output rate and shot size are operator controlled. Designed with a vacuum drawback to prevent very thin materials from dripping and reduce tailing or stringing with thicker materials.
Applications include solder paste, silicone, oil, grease, UV adhesive, CA, anaerobic, flux.
Weight: 1.82 kg (4 lbs.)
Delivery information: Includes air hose.