Flex500™ HVC Series Flexible Round Caps, Caplugs
Supplier: Caplugs
HVC-3625-18
76245-386BX 1804.84 USD
76245-386
Flex500™ HVC Series Flexible Round Caps, Caplugs
Fittings and Connectors
The HVC series Flex 500™ vinyl caps are temperature-resistant and protect up to 500 °F. They are extremely flexible and durable protective closures which are ideal for use as thread protectors, dust and moisture seals, and act as masking caps during painting, plating or powder coating operations.