Tool Extractor, Jensen Tools
Supplier: TESTEQUITY LLC TE
EX5
76245-298EA 90.03 USD
76245-298
Tool Extractor, Jensen Tools
Puller and Inserter Tools
These extracter use to rmeove the chip, range of PLCC sizes from 20 pins through 84 pins from any socket without damage to the chip.
  • ESD safe handles for use on all static safe components
  • Made of stainless steel
Spring loaded one-hand design requires no pulling, just squeeze handles and chip is lifted from the socket.