Shoe Covers, Total Source Manufacturing
Supplier: TOTAL SOURCE MANUFACTURING TE
APP0330-RS-XL
76245-228CS 254.62 USD
76245-228
Shoe Covers, Total Source Manufacturing
Disposable Garments
Shoe covers are made of 100% spunbond polypropylene. This non-woven fabric helps filter particulates for contamination control in critical environments.
  • Comfortable elastic ankles to fit over most shoes
  • Skid-free soles are available for greater traction and improved safety
Packaging: Packaged 100 per bag, 3 bags per case.